Order Entry
Switzerland
ContactUsLinkComponent
 
Dibutyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D)
  PROOCILDLM13671.2
 :  
Dibutyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D)
  PROOCILDLM13671.2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    93952-11-5
  • Description:
    Dibutyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D)
  • Formula:
    C₁₆H₂₂O₄
  • Description:
    Dibutyl phthalate-[ring-D4]