
5-Bromo-2′-deoxyuridine-5′-triphosphate (5-Bromo-dUTP) 10 mM in TE buffer
Catalog # 40025.
Supplier: Biotium
CAS Number:
Specifications
- Pack type:Plastic vial
- Pk:25 µl
- Seq:1166
Specifications
About this item
:
:612.99 g/mol
:GHS06
Storage Temperature:Freezer
CAS Number:102212-99-7
Formula:C₉H₁₁BrN₂Na₃O₁₄P₃
MDL Number:MFCD00057405