Order Entry
Switzerland
ContactUsLinkComponent
 
5-Bromo-2'-deoxyuridine (broxuridine) 95% (by HPLC), off-white solid
Catalog # 40024.
Supplier:  Biotium
CAS Number:  
undefined
5-Bromo-2'-deoxyuridine (broxuridine) 95% (by HPLC), off-white solid
Catalog # 40024.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU
 :
 :307.10 g/mol
Storage Temperature:Refrigerator
EINECS:200-415-9
CAS Number:59-14-3
Formula:C₉H₁₁BrN₂O₅
MDL Number:MFCD00006529