Order Entry
Switzerland
ContactUsLinkComponent
Isopropyltriphenylphosphonium iodide 98%
Irritant
Isopropyltriphenylphosphonium iodide 98%
Certificates
undefined
Irritant
Isopropyltriphenylphosphonium iodide 98%

Specification Test Results

  • Appearance:
    Light yellow to beige to light brown Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    195 to 200 °C
  • Titration Argentometric:
    ≥97.5 %
  • Specific optical rotation:
    +67° to +69° (20 °C, 589 nm) (c=2.3, CHCl3)

Specification Test Results

Related Information

Frequently Bought Together