Order Entry
Switzerland
ContactUsLinkComponent
Anthracene-[D10] 200 µg/ml in isooctane (98% D), CIL
Anthracene-[D10] 200 µg/ml in isooctane (98% D), CIL
Catalog # PROOCILDLM-102-1.2
CAS Number:  
Anthracene-[D10] 200 µg/ml in isooctane (98% D), CIL
Catalog # PROOCILDLM-102-1.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    1719-06-8
  • Description:
    Anthracene-[D10] 200 µg/ml in isooctane (98% D)
  • Formula:
    C₁₄D₁₀
  • Description:
    Anthracene-[D10]

Specifications

About this item