Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Lignoceric acid

Lignoceric acid

Irritant
Supplier:   Cayman Chemical
Lignoceric acid
Supplier:  Cayman Chemical
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  Tetracosanoic acid
  • CAS Number:557-59-5
  • Formula:C₂₄H₄₈O₂
  • MDL Number:MFCD00002810
  • Molecular Weight:368.64 g/mol
  • Merck Index:13,05509
  • Boiling Pt:406 °C (1013 hPa)
  • Melting Pt:87 °C
  • Density:0.8207 g/cm³ (100 °C)
  • Storage Temperature:Ambient
  • EINECS:209-180-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - SFr. 35.60
    SFr. 35.60
    1 * 50 mg
       SFr. 35.60

  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - SFr. 58.90
    SFr. 58.90
    1 * 100 mg
       SFr. 58.90

  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - SFr. 126.00
    SFr. 126.00
    1 * 250 mg
       SFr. 126.00

  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - SFr. 187.00
    SFr. 187.00
    1 * 500 mg
       SFr. 187.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.