Basic fuchsin
Catalog # EHERC10428000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
- Environmentally Preferable:
Specifications
About this item
Synonyms: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
:
:337.85 g/mol
Merck Index:12,05686
Melting Pt:235 °C
Density:1.22 g/cm³ (20 °C)
EINECS:211-189-6
:GHS08
Storage Temperature:Ambient
CAS Number:632-99-5
:58969-01-0
Formula:C₂₀H₂₀ClN₃
MDL Number:MFCD00012569