

Iodine trichloride (Wijs'reagent) 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:865-44-1
:
Formula:ICl₃
MDL Number:MFCD00036291
:233.26 g/mol
Merck Index:14,5020
Melting Pt:63 °C
Density:1.23 g/cm³ (20 °C)
Flash Pt:40 °C
Storage Temperature:Freezer
EINECS:212-739-8
UN:3085
ADR:5.1,II
:GHS03
:GHS05
Related Information
Frequently Bought Together