Orders LinkContactUsLinkComponent
Tray
Tray
155×95×24 mm
Catalog # TSLA73034
Tray
Catalog # TSLA73034
Tray
155×95×24 mm
Price
SFr. 380.00
Log in for availability
1 * 1 ST
Product Details & Documents
Trays made of quartz.
  • Temperature resistance up to +1050 °C
  • Highly resistant to chemicals
Specifications
  • W×D×H:
    155×95×24 mm