ContactUsLinkComponent
N,N-Dimethylacetamide, SupraSolv® for headspace gas chromatography, Supelco®
  1.00399.1000
 :  Merck
 :  
undefined
N,N-Dimethylacetamide, SupraSolv® for headspace gas chromatography, Supelco®
  1.00399.1000
 :  Merck
 :  
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
 :
 :87.12 g/mol
 :13,03254
 :166.1 °C (1013 hPa)
 :–20 °C
 :0.937 g/cm³ (20 °C)
 :70 °C
 :204-826-4
 :2810
 :6.1,III
 :M-Clarity™ quality level = MQ100
 :GHS08
 :GHS07
 :Ambient
 :127-19-5
 :CH₃CON(CH₃)₂
 :MFCD00008686