Orders LinkContactUsLinkComponent
Bandages, Askina® Elast
Elastic fixation bandage, wide-mesh, loose in the box, 4 m × 60 mm
  BRAU9048120
 :   B. Braun
Bandages, Askina® Elast
  BRAU9048120
 :  B. Braun
Bandages, Askina® Elast
Bandages, Askina® Elast
Elastic fixation bandage, wide-mesh, loose in the box, 4 m × 60 mm
SFr. 21.10
1 * 20 ST
Elastic, wide-mesh fixation bandage made from 70% viscose and 30% polyamide. For all types of fixation dressings and light support bandages.
  • Very comfortable thanks to high natural fibre content
  • Good adhesion of the individual turns of the bandage thanks to creped woven structure
  • Approx. 130% lengthways elasticity
  • Can be sterilised in an autoclave
  • Description:
    Elastic fixation bandage, wide-mesh, loose in the box
  • L×W:
    4 m × 60 mm