Order Entry
Switzerland
ContactUsLinkComponent
 
Cover glasses
  630-2746
undefined
Cover glasses
  630-2746

 

  • L×W:
    24×40 mm
  • Seq:
    09
  • Thickness:
    0,17 ±0,01 mm
  • Pk:
    1.000

 

 

Cover glasses with controlled thickness.

  • Free from colour distortion borosilicate glass D 263™ M
  • Excellent quality: Low levels of inclusions, striae, bubbles and streaks
  • High spectral transmission, excellent flatness, optimum resistance to chemicals
 : ISO 8255
 : In tight-closing plastic boxes for protection against damp and climatic influences.