Sampling spoons, sterile
Catalog # NASCB01041AWA
Supplier: NASCO FORT ATKINSON
Specifications
- Description:Sampling spoons, sterile
- Environmentally Preferable:
- Pk:100
Specifications
About this item
PS, white. These gas sterilised spoons are ideal for collecting samples.
- Made of sturdy, durable PS
- Ideal for use with Whirl-Pak® bags
The procedure for contamination-free use is to open the bag, push one spoon up, pull it out, and reclose the bag.
Packaging: 20 per bag, 100 per box.