ContactUsLinkComponent
Laboratory freezers, MediLine, LGPv series
Laboratory freezers, MediLine, LGPv series
Supplier:  Liebherr
Certificates
undefined
Laboratory freezers, MediLine, LGPv series
Supplier:  Liebherr
Specifications
  • Catalog No:
  • 472-0118
  • 472-2389
  • Model:
  • LPGv 8420
  • LGPv 1420
  • Cat. No.:
  • 472-0118
  • 472-2389
  • Air circulation:
  • Ventilated
  • Ventilated
  • Display:
  • LCD touch screen
  • LCD touch screen
  • Defrost:
  • Automatic
  • Automatic
  • Alarm:
  • Audible and visual
  • Audible and visual
  • Shelves/drawers:
  • 5 shelves
  • 8 adjustable grid shelves, plastic coated
  • Door type:
  • 1 door
  • 2 doors
  • Door lock:
  • Yes
  • Yes
  • Access ports:
  • 1 for PT100 probe (not supplied), Ø 8 mm
  • 1 for PT100 probe (not supplied), Ø 8 mm
  • Weight:
  • 197 kg
  • 262 kg
Specifications
Related Information
Product Information