Culture flasks, Erlenmeyer shape, BIOGEN
Supplier: witeg Labortechnik
Certificates
About this item
Culture flasks, made of borosilicate glass, Erlenmeyer shape, designed for orbital shakers.
- Straight neck for metal caps
- Triple baffled shake flask
- Excellent chemical and temperature resistance
Frequently Bought Together