Graduated pipettes for encymatic analysis, Class AS
Supplier: witeg Labortechnik
Certificates
About this item
Pipettes for enzymatic analysis.
- For partial delivery
- Main point ring-graduation
- Diffico-brown
- Calibrated to contain „EX"
DIN 12699
Certifications: DIN 12699. Conformity certified (except for article with *).
Frequently Bought Together