Orders LinkContactUsLinkComponent
Disposable bailers
Disposable bailers
Supplier:   Sampling Systems
Disposable bailers
Supplier:  Sampling Systems
Certificates
Disposable bailers
Disposable bailers
2 Options Available Below
Product Details & Documents
Disposable bailers designed for sampling from wells or storage silos.
Packaging: Individually bagged.
Caution: Not suitable for high density liquids.
Product Family Options
Product Information
  • Material
    Capacity
    Ø
    Length
    Availability
    Price
  • SAMP5040A-1000
    Disposable bailers
    Disposable bailers
    HDPE
    1000 ml
    40 mm
    950 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - SFr. 440.00
    SFr. 440.00
    1 * 24 ST
       SFr. 440.00
  • SAMP5040W-1000
    Weighted disposable bailers
    Weighted disposable bailers
    HDPE
    1000 ml
    40 mm
    950 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - SFr. 487.00
    SFr. 487.00
    1 * 24 ST
       SFr. 487.00