Orders LinkContactUsLinkComponent
Cup samplers
Cup samplers
Supplier:   Sampling Systems
Cup samplers
Supplier:  Sampling Systems
Certificates
Cup samplers
5 Options Available Below
Product Details & Documents
Stainless steel cup samplers designed for liquids and suspensions.
  • Single point liquid sampler with removable cup
  • Simple operation, easy to clean and hygienic design
  • Discrete samples to be taken from known heights within the product
Product Family Options
Product Information
  • Material
    Capacity
    Length
    Ø
    Availability
    Price
  • SAMP5030A-600
    Cup sampler for liquids
    Cup sampler for liquids
    Stainless steel 316
    100 ml
    600 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 548.00
    SFr. 548.00
    1 * 1 ST
       SFr. 548.00
  • SAMP5030A-1000
    Cup sampler for liquids
    Cup sampler for liquids
    Stainless steel 316
    100 ml
    1000 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 572.00
    SFr. 572.00
    1 * 1 ST
       SFr. 572.00
  • SAMP5030A-1500
    Cup sampler for liquids
    Cup sampler for liquids
    Stainless steel 316
    100 ml
    1500 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 591.00
    SFr. 591.00
    1 * 1 ST
       SFr. 591.00
  • SAMP5030J-1000
    Cup sampler, Jumbo
    Cup sampler, Jumbo
    Stainless steel 316
    500 ml
    1000 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’130.00
    SFr. 1’130.00
    1 * 1 ST
       SFr. 1’130.00
  • SAMP5030J-1500
    Cup sampler, Jumbo
    Cup sampler, Jumbo
    Stainless steel 316
    500 ml
    1500 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’150.00
    SFr. 1’150.00
    1 * 1 ST
       SFr. 1’150.00
Accessories
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0514
    Nylon
    7 - 14 mm
    Wire
    Log in for availability
    1 * 1 ST - SFr. 62.90
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0513
    Nylon
    7 - 14 mm
    Wire
    Log in for availability
    1 * 1 ST - SFr. 52.40