Orders LinkContactUsLinkComponent
Nessler cylinders
Nessler cylinders
Nessler cylinders
Nessler cylinders
Colour comparison cylinders according to Nessler for comparators, with optically plane bottom.
Product Family Options
Tall form
  • Description
  • Nessler cylinder 50 ml, tall form, with optically plane bottom, circular mark at 50 ml, Ø×L: 24×180 mm
    1 * 1 ST
    1 * 1 ST - SFr. 42.30
    SFr. 42.30
    1 * 1 ST
       SFr. 42.30
  • Nessler cylinder 100 ml, tall form, with optically plane bottom, circular mark at 100 ml, Ø×L: 32×220 mm
    1 * 1 ST
    1 * 1 ST - SFr. 45.50
    SFr. 45.50
    1 * 1 ST
       SFr. 45.50
  • Nessler cylinder 100 ml, tall form, with optically plane bottom, circular mark at 50/100 ml, Ø×L: 32×220 mm
    1 * 1 ST
    1 * 1 ST - SFr. 45.50
    SFr. 45.50
    1 * 1 ST
       SFr. 45.50
Low form
  • Description
  • Nessler cylinder 50 ml, low form, with optically plane bottom, circular mark at 50 ml, Ø×L: 26×150 mm
    1 * 1 ST
    1 * 1 ST - SFr. 42.30
    SFr. 42.30
    1 * 1 ST
       SFr. 42.30
  • Nessler cylinder 100 ml, low form, with optically plane bottom, circular mark at 100 ml, Ø×L: 34×180 mm
    1 * 1 ST
    1 * 1 ST - SFr. 45.50
    SFr. 45.50
    1 * 1 ST
       SFr. 45.50