Bottles, narrow or wide neck, square or rectangular, with thread
Supplier: BERLIN PACKAGING RIXIUS GMBH
Bottles, narrow or wide neck, square or rectangular, with thread
Supplier: BERLIN PACKAGING RIXIUS GMBH
Certificates
Jump to:
About this item
HDPE, natural.
- Rigid and shatter-proof
- Resistant to water, aqueous solutions and most chemicals
- Excellent water vapour resistance
- Ribbed sides for secure grip
Square or rectangular bottles with large filling opening for packaging and storage of liquids, powders and granulates.
Ordering information: Screw caps must be ordered seperately.
25 Options Available Below
Product Details & Documents
HDPE, natural.
- Rigid and shatter-proof
- Resistant to water, aqueous solutions and most chemicals
- Excellent water vapour resistance
- Ribbed sides for secure grip
Square or rectangular bottles with large filling opening for packaging and storage of liquids, powders and granulates.
Ordering information: Screw caps must be ordered seperately.
Documents
RIXI2-E0-1000-40_TDS_EN
Datasheet
Product Family Options
Narrow neck, rectangular
Product Information
- CapacityNeck I-ØW×D×HAvailabilityPrice
Wide neck, rectangular
Product Information
- CapacityNeck I-ØW×D×HAvailabilityPrice
Wide neck, square
Product Information
- CapacityNeck I-ØW×D×HAvailabilityPrice