ContactUsLinkComponent
Splash heads, bent, acc. to Stutzer
Splash heads, bent, acc. to Stutzer
Catalog # WITG1.425.005
Supplier:  witeg Labortechnik
undefined
Splash heads, bent, acc. to Stutzer
Catalog # WITG1.425.005
Supplier:  witeg Labortechnik
Specifications
  • Cone size:
    24/29 NS
  • Pk:
    1
Specifications
About this item
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Two standard ground cones, inclined at an angle of 75°. Placed directly on the distillation flask to intercept droplets.