ContactUsLinkComponent
Splash heads, acc. to Stutzer, straight
Splash heads, acc. to Stutzer, straight
Catalog # WITG1.420.004
Supplier:  witeg Labortechnik
undefined
Splash heads, acc. to Stutzer, straight
Catalog # WITG1.420.004
Supplier:  witeg Labortechnik
Specifications
  • Cone size:
    29/32 NS
  • Socket size:
    29/32 NS
  • Pk:
    1
Specifications
About this item
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Placed directly on the distillation flask to intercept droplets, with two standard ground joints.