Freeze dry systems using dry ice, 12 ports
Catalog # LBCP7522700
Supplier: LABCONCO
Specifications
- Ice capacity (l):1 L
- Refrigerant:Dry Ice
- Dimensions:Ø×H 248×222 mm (Condenser)
- Type:Bench
- Weight:5 kg
- Condenser temperature:−75 °C
- Condenser material:Stainless steel
- Pk:1
Specifications
About this item
Benchtop freeze drying system, type 304 stainless steel with 12 ports, -75 °C. Ideal for the laboratory with occasional lyophilisation requirements, the Dry Ice Benchtop Freeze Dry System provides a simple, economical means of freeze-drying. It is equipped with a center well for dry ice and solvent that serves as a water vapour collector and doubles as a convenient pre-freezing bath. Flasks, serum bottles and ampules may be frozen by dipping and rotating them in the well. Chamber size 248 x222 mm
- Ideal for processing materials with low eutectic points
- Includes dry ice/solvent center well with 1,92 litre capacity and cover
- Twelve valves and single run capacity of 1 litre
- Valves accommodate either 1/2" or 3/4" flask adapters
- 1/2" outside diameter port for connection to vacuum pump and 3/8" outside diameter port or connection to vacuum gauge
Ordering information: Supplied without rotary vane direct drive vacuum pump (requires minimum displacement of 144 l/min, 0,002 mbar ultimate pressure and fitting suitable for 3/4" ID vacuum hose). Also requires drying accessory: chamber, manifold or tray dryer (some models include a stoppering tray dryer), freeze dry glassware if not bulk freeze drying, a vacuum gauge and tubing.