ContactUsLinkComponent
Size exclusion and ligand exchange chromatography columns, SUGAR SC
Size exclusion and ligand exchange chromatography columns, SUGAR SC
Supplier:  Resonac
Certificates
Size exclusion and ligand exchange chromatography columns, SUGAR SC
Supplier:  Resonac
Specifications
  • Catalog No:
  • 554-2226
  • 554-2231
  • 554-2223
  • Description:
  • EP SC1011-7F
  • SUGAR SC-G 6B
  • SUGAR SC1011
  • Max. pressure:
  • 2,5 MPa (25 bar)
  • 4,0 MPa (40 bar)
  • pH range:
  • 3 - 7
  • 3 - 7
  • 3 - 7
  • Temperature limit:
  • 70 - 85 (≤95 °C)
  • 70 - 85 (≤95 °C)
  • 70 - 85 (≤95 °C)
  • USP code:
  • L19, L22
  • L19, L22
  • L19, L22