ContactUsLinkComponent
Ion exclusion chromatography columns, SUGAR SH
Ion exclusion chromatography columns, SUGAR SH
Supplier:  Resonac
Certificates
Ion exclusion chromatography columns, SUGAR SH
Supplier:  Resonac
Specifications
  • Catalog No:
  • 554-2229
  • 554-2222
  • 554-2221
  • Description:
  • SUGAR SH-G
  • SUGAR SH1821
  • SUGAR SH1011
  • Max. pressure:
  • 5,0 MPa (50 bar)
  • 5,0 MPa (50 bar)
  • Temperature limit:
  • 50 - 60 (≤95 °C)
  • 50 - 60 (≤95 °C)
  • 50 - 60 (≤95 °C)
  • USP code:
  • L17, L22
  • L17, L22
  • L17, L22