Order Entry
Switzerland
ContactUsLinkComponent
Sudan black B, Sigma-Aldrich®
Sudan black B, Sigma-Aldrich®
Catalog # 199664-25G
CAS Number:  
undefined
Sudan black B, Sigma-Aldrich®
Catalog # 199664-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :456.55 g/mol
Storage Temperature:Ambient
EINECS:224-087-1
CAS Number:4197-25-5
Formula:C₂₉H₂₄N₆
MDL Number:MFCD00006919