Methotrexate
Glass bottle for solids, 500 mg
Catalog # PROOMM0040.00
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
- CAS Number:59-05-2
- Formula:C₂₀H₂₂N₈O₅
- MDL Number:MFCD00150847
- Molecular Weight:454.44 g/mol
- Storage Temperature:Freezer
- EINECS:200-413-8
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



