Methotrexate
Glass bottle for solids, 100 mg
Catalog # PROOICRS0345
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
- CAS Number:59-05-2
- Formula:C₂₀H₂₂N₈O₅
- MDL Number:MFCD00150847
- Molecular Weight:454.44 g/mol
- Storage Temperature:Freezer
- EINECS:200-413-8
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



