Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Pepstatin A
Pepstatin A
Pepstatin A
Catalog # CHMCEI9
Supplier:
Chemicon
CAS Number:
Bild vergrößern
Pepstatin A
Catalog # CHMCEI9
Supplier:
Chemicon
CAS Number:
Specifications
Pk:
100 mg
CAS No.:
26305-03-3
Storage conditions:
Refrigerator
MDL No.:
MFCD00060740
Protein/peptide name:
Pepstatin A
Molecular formula:
C₃₄H₆₃N₅O₉
Specifications
About this item
Produktdetails
Produktdetails