Pepstatin A
Catalog # CHMCEI10
Supplier: Chemicon
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- CAS No.:26305-03-3
- Storage conditions:Refrigerator
- MDL No.:MFCD00060740
- Protein/peptide name:Pepstatin A
- Molecular formula:C₃₄H₆₃N₅O₉