VWR®, Pepstatin A
Catalog # J583-25MG
Anbieter: VWR Chemicals
CAS-Nummer:
Technische Daten
- Pack type:Amber bottle
- Pk:25 mg
- CAS No.:26305-03-3
- Seq:02
- Protein/peptide name:Pepstatin A
- Grade:Ultra Pure Grade
- Molecular formula:C₃₄H₆₃N₅O₉
Technische Daten
Über diesen Artikel
An aspartic acid protease inhibitor of pepsin, renin, and cathepsin D. Also an inhibitor of the HIV protease. Not water soluble.