Actinomycin D
Glass bottle for solids, 25 mg
Catalog # MOLE32117981-25MG
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:50-76-0
- Formula:C₆₂H₈₆N₁₂O₁₆
- MDL Number:MFCD00005033
- Molecular Weight:1255.5 g/mol
- UN:3462
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



