Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Heim
Proteins and Peptides
E-64C
E-64C
BACE4027912.0001
:
BACHEM BIOCHEMICA
:
E-64C
BACE4027912.0001
:
BACHEM BIOCHEMICA
:
Pack type:
Glass bottle for solids
Pk:
1 mg
CAS No.:
76684-89-4
Protein synonyms:
3S)-(+)-trans-Epoxy succinyl-L-leucylamido-3-methyl butane)|((2S
Protein/peptide name:
E-64C