Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Amphotericin B

Amphotericin B

Glass bottle for solids, 1 g
Catalog # A1907.0001
Irritant
Supplier:   PanReac AppliChem
Amphotericin B
Catalog # A1907.0001
Supplier:  PanReac AppliChem
CAS Number:  1397-89-3
Amphotericin B

Glass bottle for solids, 1 g

CAS Number:  1397-89-3
Price
SFr. 476.00
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
  • CAS Number:1397-89-3
  • Formula:C₄₇H₇₃NO₁₇
  • MDL Number:MFCD00877763
  • Molecular Weight:924.08 g/mol
  • Storage Temperature:Freezer
  • EINECS:215-742-2

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.