Thymidine (2'-deoxythymidine)
Glass bottle for solids, 1 g
Catalog # ROTH3005.1
Marketsource
Supplier: Roth Carl
Thymidine (2'-deoxythymidine)
Catalog # ROTH3005.1
Supplier: Roth Carl
Marketsource
CAS Number: 50-89-5
Jump to:
Product Details & Documents
Synonyms: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
- CAS Number:50-89-5
- Formula:C₁₀H₁₄N₂O₅
- MDL Number:MFCD00006537
- Molecular Weight:242.23 g/mol
- Storage Temperature:Ambient
- EINECS:200-070-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



