Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Thymidine (2'-deoxythymidine) ≥99%

Thymidine (2'-deoxythymidine) ≥99%

Health
Irritant
Thymidine (2'-deoxythymidine) ≥99%
Certificates
Thymidine (2'-deoxythymidine) ≥99%
Thymidine (2'-deoxythymidine) ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 50-89-5 Molecular Formula: C10H14N2O5 Formula Weight: 242.23 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 186-190° Beilstein Reference: 89285 Merck Reference: 14,9397 Optical Rotation: +19° (c=1 in water) MDL No.: MFCD00006537

Synonyms:  1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
  • CAS Number:50-89-5
  • Formula:C₁₀H₁₄N₂O₅
  • MDL Number:MFCD00006537
  • Molecular Weight:242.23 g/mol
  • Merck Index:14,9397
  • Melting Pt:186…190 °C
  • Storage Temperature:Ambient
  • EINECS:200-070-4
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.