Order Entry
Switzerland
ContactUsLinkComponent
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate
Catalog # MOLEM79277079
Supplier:  Molekula
CAS Number:  
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate
Catalog # MOLEM79277079
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :391.2 g/mol (anhy)
Storage Temperature:Refrigerator
EINECS:224-961-2
CAS Number:4578-31-8
Formula:C₁₀H₁₂N₅Na₂O₇P·H₂O
MDL Number:MFCD00065023