Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Puromycin aminonucleoside
Puromycin aminonucleoside
Puromycin aminonucleoside
Puromycin aminonucleoside
Catalog # CHMCAB3258
Supplier:
Chemicon
CAS Number:
Expand image
Puromycin aminonucleoside
Catalog # CHMCAB3258
Supplier:
Chemicon
CAS Number:
Specifications
Pk:
50 µl
Seq:
1202
Specifications
About this item
Product Details
:
:
294.31 g/mol
EINECS:
200-388-3
CAS Number:
58-60-6
Formula:
C₁₂H₁₈N₆O₃
MDL Number:
MFCD00063462
Product Details