Order Entry
Switzerland
ContactUsLinkComponent
 
Irritant
Naphthol AS-MX phosphate, Sigma-Aldrich®
Catalog # 855-20ML
CAS Number:  
Irritant
Naphthol AS-MX phosphate, Sigma-Aldrich®
Catalog # 855-20ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    20 mL
  • Seq:
    2305

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ500 (more info.)
EINECS:216-480-1
 :GHS07
Storage Temperature:Freezer
CAS Number:1596-56-1
Formula:C₁₉H₁₈NO₅P
MDL Number:MFCD00042711