Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Auramine O

Toxic
Marketsource
Supplier:   Roth Carl
Auramine O
Supplier:  Roth Carl
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Pyoctanunum aureum, 4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Pyoktanin Yellow, 4,4'-(Iminomethylene)bis(N,N-dimethylaniline) hydrochloride, Basic Yellow 2, Canary Yellow
  • CAS Number:2465-27-2
  • Formula:C₁₇H₂₂ClN₃
  • MDL Number:MFCD00012484
  • Molecular Weight:303.84 g/mol
  • Storage Temperature:Ambient
  • EINECS:219-567-2
  • UN:2811
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 24.60
    SFr. 24.60
    1 * 25 g
       SFr. 24.60

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - SFr. 43.10
    SFr. 43.10
    1 * 50 g
       SFr. 43.10

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.