1-Naphthyl phosphate monosodium salt monohydrate, Millipore®
Catalog # 479775-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: 1-Naphthylphosphoric Acid Monosodium Salt Monohydrate, Monosodium 1-Naphthyl Phosphate Monohydrate, Sodium 1-Naphthyl Hydrogen Phosphate Monohydrate, Phosphoric Acid 1-Naphthyl Ester Monosodium Salt Monohydrate
:
:264.15 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:81012-89-7
Formula:C₁₀H₈NaO₄P·1H₂O
MDL Number:MFCD00150615