Order Entry
Switzerland
ContactUsLinkComponent
 
Health
Carbenicillin disodium salt
Catalog # MSPPC0109.0025
CAS Number:  
Health
Carbenicillin disodium salt
Catalog # MSPPC0109.0025
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :422.363 g/mol
EINECS:225-360-8
 :GHS08
Storage Temperature:Refrigerator
CAS Number:4800-94-6
Formula:C₁₇H₁₆N₂O₆S·2Na
MDL Number:MFCD00077683