Order Entry
Switzerland
ContactUsLinkComponent
Hydroxyethyl cellulose, M.W. ~1000000, 1500 - 2500 cps (1%, 25 °C)
Hydroxyethyl cellulose, M.W. ~1000000, 1500 - 2500 cps (1%, 25 °C)
Catalog # PROOFIP20
CAS Number:  
Hydroxyethyl cellulose, M.W. ~1000000, 1500 - 2500 cps (1%, 25 °C)
Catalog # PROOFIP20
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:9004-62-0
MDL Number:MFCD00072770