Nonidet® P 40 Substitute (NP-40)
Catalog # EURLEMR181500
Supplier: EUROCLONE
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
:
EINECS:500-024-6
:GHS07
Storage Temperature:Ambient
CAS Number:9016-45-9
MDL Number:MFCD00132851