Order Entry
Switzerland
ContactUsLinkComponent
Phosphorus standard solution, 1,000 mg/L P in water (from (NH₄)PO₄H₂) standard for AAS and ICP
Phosphorus standard solution, 1,000 mg/L P in water (from (NH₄)PO₄H₂) standard for AAS and ICP
Catalog # PERCN9300139
Supplier:  PerkinElmer
CAS Number:  
Phosphorus standard solution, 1,000 mg/L P in water (from (NH₄)PO₄H₂) standard for AAS and ICP
Catalog # PERCN9300139
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    1
  • Pk Info:
    1× 500 ml
  • Element:
    Phosphorus
  • Symbol:
    P

Specifications

About this item