Order Entry
Switzerland
ContactUsLinkComponent
Lanthanum standard solution, 1000 mg/L La in dil. nitric acid (from La₂O₃) standard for AAS and ICP
Lanthanum standard solution, 1000 mg/L La in dil. nitric acid (from La₂O₃) standard for AAS and ICP
Catalog # PERCN9300127
Supplier:  PerkinElmer
CAS Number:  
Lanthanum standard solution, 1000 mg/L La in dil. nitric acid (from La₂O₃) standard for AAS and ICP
Catalog # PERCN9300127
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Lanthanum
  • Symbol:
    La

Specifications

About this item