
AMP (2-Amino-2-methylpropanol), high purity
0136-4L
:
- Pack type:Plastic bottle
- Pk:4 L
- Seq:5162
- pH (0.1 M, Water) @25 °C:11.0 - 12.0
- Purity (anhydrous):98.5 %
- Reassay Date:REPORT
:
:13,00448
:165 °C (1013 hPa)
:18…26 °C
:0.667 g/cm³ (20 °C)
:67 °C
:89.14 g/mol
:Ambient
:204-709-8
:GHS07
:124-68-5
:(CH₃)₂C(NH₂)CH₂OH
:MFCD00008051