ContactUsLinkComponent
Rotor for Cryofuge™ 6000i/8500i series
  521-1384
undefined
Rotor for Cryofuge™ 6000i/8500i series
  521-1384
  • Description:
    Windshielded rotor with lid, without buckets, 6×1000 ml, 5050 min⁻¹ with Cryofuge 6000i and Cryofuge 8500i
  • Seq:
    01
  • Environmentally Preferable:
  • Pk:
    1
Windshielded rotor for the large capacity centrifuges Cryofuge 6000i and 8500i.
  • Swinging bucket
  • Rotor body made of stainless steel; bucket made of aluminium
  • Comes with a lid but without buckets