Slot samplers, SteriWare®
Supplier: Sampling Systems
Certificates
About this item
These cross sectional multilevel disposable powder samplers are designed for powders. The robust HDPE construction make this to be used in huge number of testing applications.
- BSE/TSE-free
- Full batch traceability
- Suitable for use in ATEX zones 0, 1 and 2
- Available as gamma irradiated sterile (SAL 10-6)
Can take a composite from the top, middle and bottom of the powder bed – Moulded and packed in a cleanroom to ensure cleanliness.
Ideal for taking a multilevel, cross sectional sample from powders and granules. Insert the sampler and rotate the handle to take a representative sample.
Certifications: FDA, EU 10/2011 and EC 1935/2004.
Packaging: Individually bagged in a heat-sealed PE bag.