Orders LinkContactUsLinkComponent
Square HDPE Bottle
Square HDPE Bottle
Supplier:   Sampling Systems
Square HDPE Bottle
Supplier:  Sampling Systems
Certificates
Square HDPE Bottle
Square HDPE Bottle
3 Options Available Below
Product Details & Documents
Square HDPE bottles for use with a range of liquids and powders.
  • Different sizes available
  • Ribbed sides for easy handling
  • Available from stock
  • Fast delivery
This range of HDPE containers are perfect for use with a range of powders and liquids. They have a handy square space-saving shape and incorporate side ridges for easy handling when wet.
Product Family Options
Product Information
  • Capacity
    Material
    Availability
    Price
  • SAMP8165A-250
    Square bottle
    Square bottle
    250 ml
    HDPE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 7.20
    SFr. 7.20
    1 * 1 ST
       SFr. 7.20
  • SAMP8165A-500
    Square bottle
    Square bottle
    500 ml
    HDPE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 8.90
    SFr. 8.90
    1 * 1 ST
       SFr. 8.90
  • SAMP8165A-1000
    Square bottle
    Square bottle
    1000 ml
    HDPE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 9.80
    SFr. 9.80
    1 * 1 ST
       SFr. 9.80
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..